Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXL2
Entrez Gene ID
100622956
Gene Name
forkhead box L2
Gene Aliases
-
Chromosome Location
Chr.13: 79708693 - 79709825 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244665.1 NP_001231594.1 1 - 1 217 103

View Full Product Details