Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN12
Entrez Gene ID
100512202
Gene Name
tetraspanin 12
Gene Aliases
-
Chromosome Location
Chr.18: 26046645 - 26112924 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244673.1 NP_001231602.1 3 - 4 373 75
XM_013985637.2 XP_013841091.1 - 431 75
XM_013985638.2 XP_013841092.1 - 435 75
XM_021078478.1 XP_020934137.1 - 469 75
XM_021078479.1 XP_020934138.1 - 741 75
XM_021078480.1 XP_020934139.1 - 376 75
GenBank mRNA AK233373.1 - 3 - 4 373 75

View Full Product Details