Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNFN
Entrez Gene ID
100512661
Gene Name
cornifelin
Gene Aliases
-
Chromosome Location
Chr.6: 49568224 - 49574070 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244689.1 NP_001231618.1 2 - 3 199 68
XM_005655861.3 XP_005655918.1 - 319 68
XM_013988596.2 XP_013844050.1 - 1995 68
XM_021093304.1 XP_020948963.1 - 2133 68
XM_021093305.1 XP_020948964.1 - 243 68
XM_021093306.1 XP_020948965.1 - 215 68
XM_021093307.1 XP_020948966.1 - 200 68
XM_021093308.1 XP_020948967.1 - 215 68
XM_021093309.1 XP_020948968.1 - 189 68
XM_021093310.1 XP_020948969.1 - 280 68
GenBank mRNA AK349949.1 - 2 - 3 199 68

View Full Product Details