Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBLN2
Entrez Gene ID
100517134
Gene Name
cerebellin 2 precursor
Gene Aliases
-
Chromosome Location
Chr.1: 150966003 - 150980757 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244702.1 NP_001231631.1 4 - 5 878 64
XM_021073355.1 XP_020929014.1 - 2909 64
XM_021073411.1 XP_020929070.1 - 852 64
GenBank mRNA AK392456.1 - 2 - 3 323 64

View Full Product Details