Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIIP
Entrez Gene ID
100512719
Gene Name
migration and invasion inhibitory protein
Gene Aliases
-
Chromosome Location
Chr.6: 72064634 - 72074981 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244704.1 NP_001231633.1 4 - 5 796 72
XM_005665007.3 XP_005665064.1 - 699 72
XM_005665008.3 XP_005665065.1 - 794 72
XM_021093311.1 XP_020948970.1 - 699 72
GenBank mRNA AK237551.1 - 4 - 5 796 72
AK347476.1 - 4 - 5 787 72
AK397949.1 - 4 - 5 784 72

View Full Product Details