Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP23
Entrez Gene ID
100512755
Gene Name
synaptosome associated protein 23
Gene Aliases
-
Chromosome Location
Chr.1: 128849465 - 128888972 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244718.1 NP_001231647.1 7 - 8 605 85
NM_001244719.1 NP_001231648.1 6 - 7 566 85
XM_005659700.3 XP_005659757.1 - 741 85
XM_013993108.2 XP_013848562.1 - 616 85
XM_013993109.2 XP_013848563.1 - 802 85
XM_013993110.2 XP_013848564.1 - 635 85
XM_013993111.2 XP_013848565.1 - 795 85
XM_013993112.2 XP_013848566.1 - 644 85
XM_021069852.1 XP_020925511.1 - 732 85
XM_021070075.1 XP_020925734.1 - 602 85
XM_021070141.1 XP_020925800.1 - 475 85
GenBank mRNA AK230671.1 - 7 - 8 605 85
AK230848.1 - 6 - 7 546 85
AK389714.1 - 6 - 7 552 85
AY609458.1 - 6 - 7 566 85

View Full Product Details