Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRIX1
Entrez Gene ID
100513502
Gene Name
biogenesis of ribosomes BRX1
Gene Aliases
-
Chromosome Location
Chr.16: 20512281 - 20520579 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244824.1 NP_001231753.1 4 - 5 424 117
GenBank mRNA AK237113.1 - 4 - 5 424 117
AY610198.1 - 4 - 5 407 117

View Full Product Details