Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5PD
Entrez Gene ID
100512624
Gene Name
ATP synthase peripheral stalk subunit d
Gene Aliases
ATP5H
Chromosome Location
Chr.12: 6241467 - 6247344 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244684.1 NP_001231613.1 1 - 2 135 100
XM_021066093.1 XP_020921752.1 - 285 100
XM_021066094.1 XP_020921753.1 - 285 100
GenBank mRNA AK231326.1 - 2 - 3 160 100
AK391854.1 - 2 - 3 175 100
AY610267.1 - 2 - 3 149 100

View Full Product Details