Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR15B
- Entrez Gene ID
-
100170411
- Gene Name
- microRNA mir-15b
- Gene Aliases
-
MIRN15B, ssc-mir-15b
- Chromosome Location
-
Chr.13: 100083172 - 100083269 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031533.2 |
|
1 - 1 |
37 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
intraflagellar transport 80 |
IFT80 |
|
-5.0kb
|
Anti-sense |
Chr. - 99943203 - 100078148 on Build Sscrofa11.1 |
Downstream |
microRNA mir-16-1 |
MIR16-1 |
MIR16-2, ssc-mir-16-1, ssc-mir-16-2 |
60.0
|
Sense |
Chr. - 100083329 - 100083405 on Build Sscrofa11.1 |
View Full Product Details