Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Prx1
Entrez Gene ID
606328
Gene Name
peroxiredoxin Q
Gene Aliases
TaPrx1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY789643.1 - - 555 70

View Full Product Details