Gene Transcripts

Gene Symbol
SMAD5
Entrez Gene ID
4090
Gene Name
SMAD family member 5
Gene Aliases
DWFC, JV5-1, MADH5
Chromosome Location
Chr. 5: 136132845 - 136182734 on Build GRCh38
UniGene ID
Hs.167700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001419.2 NP_001001419.1 7 1385
NM_001001420.2 NP_001001420.1 5 1226
NM_005903.6 NP_005894.3 6 1301
XM_017009470.1 XP_016864959.1 7 1517
GenBank mRNA AF009678.1 - 4 953
AF010601.1 - 4 1070
AK313397.1 - 6 1301
BC009682.2 - 4 1107 3906006
BX648328.1 - 6 1240
CR457424.1 - 4 939
CR749366.1 - 6 1240
CR749473.1 - 6 1228
U59913.1 - 4 1069
U73825.1 - 4 1088

View Full Product Details