Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VVSUC11
Entrez Gene ID
100232844
Gene Name
sucrose transporter-like
Gene Aliases
GSVIVT00015035001
Chromosome Location
Chr.18: 6598543 - 6607133 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF021808.1 - 2 - 3 1263 101
AF182445.1 - 2 - 3 1233 101

View Full Product Details