Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VVSUC27
Entrez Gene ID
100232846
Gene Name
sucrose transporter-like
Gene Aliases
GSVIVT00002302001, SUC27
Chromosome Location
Chr.Un_NW_002239610: 909524 - 912065 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF021810.1 - 3 - 4 1455 83

View Full Product Details