Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADH3
Entrez Gene ID
100233102
Gene Name
alcohol dehydrogenase 3
Gene Aliases
GSVIVT01010027001
Chromosome Location
Chr.18: 13409070 - 13412228 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF194175.1 - 1 - 2 69 61

View Full Product Details