Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPEPC
Entrez Gene ID
100233103
Gene Name
PEP carboxylase
Gene Aliases
GSVIVT00018735001
Chromosome Location
Chr.12: 2930226 - 2937192 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF236126.1 - 2 - 3 723 74

View Full Product Details