Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP24
Entrez Gene ID
100232887
Gene Name
metallothionein-like protein-like
Gene Aliases
GSVIVT01014340001
Chromosome Location
Chr.19: 2786796 - 2805250 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ237990.1 - 1 - 2 106 59

View Full Product Details