Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIP58
Entrez Gene ID
100233110
Gene Name
ripening-related protein-like
Gene Aliases
GSVIVT00036026001
Chromosome Location
Chr.3: 991507 - 1000138 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ237993.1 - 2 - 3 261 56

View Full Product Details