Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOR
Entrez Gene ID
100233111
Gene Name
shaker-like potassium channel
Gene Aliases
GSVIVT00032933001
Chromosome Location
Chr.14: 7381180 - 7409165 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ490336.1 - 6 - 7 1179 89

View Full Product Details