Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QOR
Entrez Gene ID
100232919
Gene Name
quinone reductase-like
Gene Aliases
GSVIVT00027476001
Chromosome Location
Chr.19: 4904750 - 4909739 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY159559.1 - 1 - 2 262 73

View Full Product Details