Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OXC
Entrez Gene ID
100232933
Gene Name
oxalyl-CoA decarboxylase-like
Gene Aliases
-
Chromosome Location
Chr.Un_NW_002239379: 368611 - 372660 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY233204.1 - 1 - 2 177 75

View Full Product Details