Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHCA1
Entrez Gene ID
100232934
Gene Name
chlorophyll a/b-binding protein
Gene Aliases
GSVIVT00016319001
Chromosome Location
Chr.17: 7001287 - 7002634 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY233205.1 - 1 - 2 148 66

View Full Product Details