Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZEP
Entrez Gene ID
100232944
Gene Name
zeaxanthin epoxidase
Gene Aliases
GSVIVT00019494001
Chromosome Location
Chr.7: 11918624 - 11926467 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY337615.1 - 6 - 7 1183 89

View Full Product Details