Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HT4
Entrez Gene ID
100232949
Gene Name
hexose transporter-like
Gene Aliases
GSVIVT00030854001
Chromosome Location
Chr.Un_NW_002239896: 1645443 - 1649004 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY538260.1 - 2 - 3 483 81

View Full Product Details