Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HT5
Entrez Gene ID
100232950
Gene Name
hexose transporter
Gene Aliases
GSVIVT00019956001
Chromosome Location
Chr.5: 4760126 - 4764839 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY538261.1 - 3 - 4 630 80

View Full Product Details