Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWINV
Entrez Gene ID
100232951
Gene Name
cell wall apoplastic invertase
Gene Aliases
-
Chromosome Location
Chr.9: 2103328 - 2107257 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY538262.1 - 3 - 4 1071 65

View Full Product Details