Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VFUL-L
Entrez Gene ID
100232953
Gene Name
FUL-like protein
Gene Aliases
FUL, GSVIVT00038079001
Chromosome Location
Chr.14: 12518771 - 12539672 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY538747.1 - 8 - 9 727 72

View Full Product Details