Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HGGT
Entrez Gene ID
100232967
Gene Name
homogentisate geranylgeranyl transferase
Gene Aliases
GSVIVT01029247001
Chromosome Location
Chr.11: 12208609 - 12218151 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY826350.1 - 10 - 11 981 79

View Full Product Details