Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GS
- Entrez Gene ID
-
100266191
- Gene Name
- glutamine synthetase
- Gene Aliases
-
1, GS1, GS1-1, GSVIVT00017890001
- Chromosome Location
-
Chr.17: 1473587 - 1477294 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X94320.1 |
- |
1 - 2 |
164 |
70 |
View Full Product Details