Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VVGT6
Entrez Gene ID
100244489
Gene Name
UDP-sugar flavonoid glycosyltransferase
Gene Aliases
-
Chromosome Location
Chr.11: 13521403 - 13526456 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB499075.1 - 1 - 2 489 64

View Full Product Details