Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP3
Entrez Gene ID
100233029
Gene Name
flowering-related B-class MADS-box protein APETALA3
Gene Aliases
GSVIVT00014506001
Chromosome Location
Chr.18: 11366348 - 11372377 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EF418603.1 - 1 - 2 189 99
GU133632.1 - 1 - 2 99

View Full Product Details