Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYB24
Entrez Gene ID
100233097
Gene Name
R2R3 Myb24 transcription factor
Gene Aliases
GSVIVT00020719001
Chromosome Location
Chr.14: 16764214 - 16768191 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU181426.1 - 2 - 3 270 98

View Full Product Details