Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RD22-C
Entrez Gene ID
100264522
Gene Name
rd22-c protein
Gene Aliases
-
Chromosome Location
Chr.4: 3265590 - 3286814 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ869894.2 - 2 - 3 191 83

View Full Product Details