Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIPK12
Entrez Gene ID
100254007
Gene Name
CBL-interacting protein kinase 12
Gene Aliases
GSVIVT00026148001
Chromosome Location
Chr.15: 7464177 - 7468960 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901240.1 - 6 - 7 551 100

View Full Product Details