Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIPK14
Entrez Gene ID
100259887
Gene Name
CBL-interacting protein kinase 14
Gene Aliases
GSVIVT00031453001
Chromosome Location
Chr.11: 13605515 - 13608929 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901242.1 - 14 - 15 1343 114

View Full Product Details