Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CIPK14
- Entrez Gene ID
-
100259887
- Gene Name
- CBL-interacting protein kinase 14
- Gene Aliases
-
GSVIVT00031453001
- Chromosome Location
-
Chr.11: 13605515 - 13608929 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FJ901242.1 |
- |
9 - 10 |
874 |
113 |
- Gene Symbol
- CIPK06
- Entrez Gene ID
-
100379632
- Gene Name
- CBL-interacting protein kinase 06
- Gene Aliases
-
VITISV_040990
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FJ901234.1 |
- |
7 - 8 |
|
113 |
View Full Product Details