Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP61
Entrez Gene ID
100232890
Gene Name
ripening-related protein-like
Gene Aliases
GSVIVT00030517001
Chromosome Location
Chr.1: 4755334 - 4756587 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ237994.1 - 1 - 2 244 98

View Full Product Details