The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002902.1 | NP_001002902.1 | 6 - 7 | 476 | 100 |
XM_012956694.2 | XP_012812148.1 | 7 - 8 | 685 | 100 | |
XM_018091156.1 | XP_017946645.1 | 6 - 7 | 523 | 100 | |
GenBank mRNA | BC074572.1 | - | 6 - 7 | 476 | 100 |
CR762304.2 | - | 5 - 6 | 427 | 100 |