Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stk35
Entrez Gene ID
447998
Gene Name
serine/threonine kinase 35
Gene Aliases
clik1, stk35l1
Chromosome Location
Chr.Un|NW_016683402: 588659 - 593480 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004779.1 NP_001004779.1 1 - 2 651 93
GenBank mRNA BC074513.1 - 1 - 2 651 93
CR761751.2 - 1 - 2 623 93

View Full Product Details