Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aimp2
Entrez Gene ID
448040
Gene Name
aminoacyl tRNA synthetase complex-interacting multifunctional protein 2
Gene Aliases
jtv1, pro0992
Chromosome Location
Chr.9: 19725733 - 19738789 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004804.1 NP_001004804.1 1 - 2 140 57
GenBank mRNA BC074561.1 - 1 - 2 140 57
CR760281.2 - 1 - 2 129 57

View Full Product Details