Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arpin
Entrez Gene ID
448248
Gene Name
actin related protein 2/3 complex inhibitor
Gene Aliases
c15orf38
Chromosome Location
Chr.3: 98561622 - 98570873 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004898.1 NP_001004898.1 5 - 6 711 76
XM_012958820.2 XP_012814274.1 6 - 7 864 76
XM_012958821.2 XP_012814275.1 6 - 7 853 76
XM_012958822.2 XP_012814276.1 5 - 6 682 76
XM_012958823.2 XP_012814277.1 5 - 6 674 76
GenBank mRNA BC075312.1 - 5 - 6 711 76

View Full Product Details