Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdzd11
Entrez Gene ID
448266
Gene Name
PDZ domain containing 11
Gene Aliases
-
Chromosome Location
Chr.8: 17760373 - 17766399 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004906.1 NP_001004906.1 3 - 4 279 65
XM_018096199.1 XP_017951688.1 2 - 3 259 65
GenBank mRNA BC075334.1 - 3 - 4 279 65
CR760601.2 - 3 - 4 287 65

View Full Product Details