Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
bicdl2
Entrez Gene ID
448330
Gene Name
BICD family like cargo adaptor 2
Gene Aliases
BICDR-2, bicdr2, ccdc64b
Chromosome Location
Chr.9: 6887205 - 6920612 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004934.1 NP_001004934.1 - 1305 90
GenBank mRNA BC075409.1 - - 1305 90

View Full Product Details