Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl10
Entrez Gene ID
448389
Gene Name
ribosomal protein L10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683541: 20092 - 24507 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004965.2 NP_001004965.1 4 - 5 347 89
XM_012954243.1 XP_012809697.1 5 - 6 386 89
GenBank mRNA BC075477.1 - 4 - 5 351 89
BC158397.1 - 4 - 5 361 89
CU025127.1 - 4 - 5 346 89

View Full Product Details