Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gadd45g
Entrez Gene ID
448442
Gene Name
growth arrest and DNA damage inducible gamma
Gene Aliases
XGadd45-gamma, cr6, ddit2, gadd45gamma, grp17, xgadd45g
Chromosome Location
Chr.1: 125856172 - 125858086 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004986.1 NP_001004986.1 2 - 3 266 77
GenBank mRNA BC075545.1 - 2 - 3 266 77
CR761710.2 - 2 - 3 261 77

View Full Product Details