Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncbp3
Entrez Gene ID
448518
Gene Name
nuclear cap binding subunit 3
Gene Aliases
c17orf85
Chromosome Location
Chr.2: 44158987 - 44184281 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005015.1 NP_001005015.1 2 - 3 299 72
GenBank mRNA BC076682.1 - 2 - 3 299 72

View Full Product Details