Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c19orf47
Entrez Gene ID
448521
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.8: 56784568 - 56795053 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005016.1 NP_001005016.1 3 - 4 217 92
XM_018096214.1 XP_017951703.1 3 - 4 404 92
GenBank mRNA BC076686.1 - 3 - 4 217 92
CR848118.2 - 3 - 4 162 92

View Full Product Details