Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt3a2
Entrez Gene ID
448540
Gene Name
UDP glycosyltransferase 3 family, polypeptide A2
Gene Aliases
-
Chromosome Location
Chr.1: 183711306 - 183722876 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005027.3 NP_001005027.3 2 - 3 221 68
XM_012959973.2 XP_012815427.1 2 - 3 518 68
XM_018092265.1 XP_017947754.1 2 - 3 464 68
XM_018092272.1 XP_017947761.1 2 - 3 524 68
XM_018092275.1 XP_017947764.1 2 - 3 219 68
GenBank mRNA AL791968.2 - 2 - 3 221 68
BC076709.1 - 2 - 3 209 68
CU025035.1 - 2 - 3 190 68

Gene Symbol
LOC100488176
Entrez Gene ID
100488176
Gene Name
UDP-glucuronosyltransferase 3A1-like
Gene Aliases
-
Chromosome Location
Chr.1: 183697145 - 183710091 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004910430.3 XP_004910487.1 2 - 3 767 68
XM_018090819.1 XP_017946308.1 2 - 3 229 68

View Full Product Details