Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tada1
Entrez Gene ID
448637
Gene Name
transcriptional adaptor 1
Gene Aliases
tada1l
Chromosome Location
Chr.4: 96819471 - 96866759 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005070.1 NP_001005070.1 6 - 7 650 70
XM_018093002.1 XP_017948491.1 6 - 7 623 70
GenBank mRNA BC076979.1 - 6 - 7 650 70
CR760219.2 - 6 - 7 739 70

View Full Product Details