Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cbx7
Entrez Gene ID
448638
Gene Name
chromobox 7
Gene Aliases
-
Chromosome Location
Chr.4: 110280278 - 110302519 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005071.1 NP_001005071.1 1 - 2 620 97
XM_012960880.2 XP_012816334.1 2 - 3 363 97
XM_018093003.1 XP_017948492.1 2 - 3 377 97
XM_018093004.1 XP_017948493.1 2 - 3 474 97
XM_018093005.1 XP_017948494.1 2 - 3 677 97
XM_018093006.1 XP_017948495.1 2 - 3 335 97
GenBank mRNA BC076980.1 - 1 - 2 620 97

View Full Product Details