Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mfap1
Entrez Gene ID
496412
Gene Name
microfibril associated protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 97727745 - 97736003 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011003.1 NP_001011003.1 7 - 8 1149 117
GenBank mRNA BC082615.1 - 7 - 8 1149 117
CR762333.2 - 7 - 8 1090 117

View Full Product Details